Product Details

SNP ID
rs184680485
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:21387625 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGTCCGTATGAAAACGTGGGTGCG[C/T]GGAATGATTCTGTTAAATATACATA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MIPEPP3 PubMed Links

Gene Details

Gene
MIPEPP3
Gene Name
mitochondrial intermediate peptidase pseudogene 3
There are no transcripts associated with this gene.

Gene
ZDHHC20
Gene Name
zinc finger DHHC-type containing 20
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286638.1 354 Intron NP_001273567.1
NM_153251.3 354 Intron NP_694983.2
XM_006719795.1 354 Missense Mutation CAC,CGC H246R XP_006719858.1
XM_006719796.1 354 Missense Mutation CAC,CGC H246R XP_006719859.1
XM_006719797.1 354 Missense Mutation CAC,CGC H246R XP_006719860.1
XM_011535030.2 354 Missense Mutation CAC,CGC H98R XP_011533332.1
XM_017020525.1 354 Intron XP_016876014.1
XM_017020526.1 354 Missense Mutation CAC,CGC H167R XP_016876015.1
XM_017020527.1 354 Missense Mutation CAC,CGC H167R XP_016876016.1

View Full Product Details