Product Details

SNP ID
rs184833633
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:100149264 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGCGGGTCACGGCCTTGGGGTGGA[C/T]GCAGAGCGGCCGTAGTGAGTAGAAG
Phenotype
MIM: 610862 MIM: 616912
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DEGS2 PubMed Links

Gene Details

Gene
DEGS2
Gene Name
delta 4-desaturase, sphingolipid 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_206918.2 617 Missense Mutation ATC,GTC I177V NP_996801.2
XM_006720043.3 617 Missense Mutation ATC,GTC I141V XP_006720106.1
Gene
EVL
Gene Name
Enah/Vasp-like
There are no transcripts associated with this gene.

View Full Product Details