Product Details

SNP ID
rs184847525
Assay Type
Functionally tested
NCBI dbSNP Submissions
7
Location
Chr.1:232986288 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAGCCCAGGCTGGACCTGATGAGC[A/G]CCTCGGCCCGCTCACGGACACTCAG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NTPCR PubMed Links

Gene Details

Gene
NTPCR
Gene Name
nucleoside-triphosphatase, cancer-related
There are no transcripts associated with this gene.

Gene
PCNX2
Gene Name
pecanex homolog 2 (Drosophila)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014801.3 5843 Missense Mutation NP_055616.3
XM_006711816.3 5843 Intron XP_006711879.1
XM_011544278.2 5843 Missense Mutation XP_011542580.2
XM_017002391.1 5843 Missense Mutation XP_016857880.1
XM_017002392.1 5843 Missense Mutation XP_016857881.1

View Full Product Details