Product Details

SNP ID
rs184549175
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:69149047 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAAAACCCACCTGCTCCAAGGTAG[A/C]CTGAGAGAGGCTGTATTCCTCCAGG
Phenotype
MIM: 612508 MIM: 612504
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ABCA10 PubMed Links

Gene Details

Gene
ABCA10
Gene Name
ATP binding cassette subfamily A member 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_080282.3 5398 Missense Mutation GCT,TCT A1507S NP_525021.3
Gene
ABCA6
Gene Name
ATP binding cassette subfamily A member 6
There are no transcripts associated with this gene.

View Full Product Details