Product Details

SNP ID
hCV180531462
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:63641384 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAAAAAAAAAAAAAGAAAGAAAGA[A/C]AGAAACTAAATGAATTTCTTTGAAG
Phenotype
MIM: 300429
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ARHGEF9 PubMed Links

Gene Details

Gene
ARHGEF9
Gene Name
Cdc42 guanine nucleotide exchange factor 9
There are no transcripts associated with this gene.

View Full Product Details