Product Details

SNP ID
rs185446950
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:48138798 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTTTCAAAAGGCATTCTGCCTTCC[A/G]TGAATACTTCCCACATTAAAACACC
Phenotype
MIM: 600583 MIM: 600058
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TEC PubMed Links

Gene Details

Gene
TEC
Gene Name
tec protein tyrosine kinase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003215.2 2976 Missense Mutation ACG,ATG T560M NP_003206.2
XM_011513737.2 2976 Missense Mutation ACG,ATG T524M XP_011512039.1
XM_011513741.1 2976 Intron XP_011512043.1
XM_017008569.1 2976 Missense Mutation ACG,ATG T444M XP_016864058.1
Gene
TXK
Gene Name
TXK tyrosine kinase
There are no transcripts associated with this gene.

View Full Product Details