Product Details

SNP ID
rs185443895
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:48373519 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCTGGCCCCTGGAAATCCACAGGT[C/T]TTCTCCCTGATCGTCTTCTCCTCCC
Phenotype
MIM: 131235 MIM: 608373
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KDELR1 PubMed Links

Gene Details

Gene
KDELR1
Gene Name
KDEL endoplasmic reticulum protein retention receptor 1
There are no transcripts associated with this gene.

Gene
SYNGR4
Gene Name
synaptogyrin 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012451.3 581 Silent Mutation GTC,GTT V32V NP_036583.2
XM_005258693.2 581 Silent Mutation GTC,GTT V32V XP_005258750.1
XM_005258694.2 581 Silent Mutation GTC,GTT V32V XP_005258751.1
XM_011526686.2 581 UTR 5 XP_011524988.1
XM_017026541.1 581 UTR 5 XP_016882030.1
Gene
TMEM143
Gene Name
transmembrane protein 143
There are no transcripts associated with this gene.

View Full Product Details