Product Details

SNP ID
rs185620086
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:105492178 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCGAGGCTGGCACTGGCACAACTA[C/T]CTGAGGATGGCTCGCAGGGCAGTCG
Phenotype
MIM: 123875
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C14orf80 PubMed Links

Gene Details

Gene
C14orf80
Gene Name
chromosome 14 open reading frame 80
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001134875.1 425 Missense Mutation CCT,TCT P100S NP_001128347.1
NM_001134876.1 425 Missense Mutation CCT,TCT P100S NP_001128348.1
NM_001134877.1 425 Missense Mutation CCT,TCT P59S NP_001128349.1
NM_001198983.1 425 Missense Mutation CCT,TCT P61S NP_001185912.1
Gene
CRIP1
Gene Name
cysteine rich protein 1
There are no transcripts associated with this gene.

View Full Product Details