Product Details

SNP ID
rs185539044
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:124962774 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCGCGAGGAGCCTGGGGCCCCCGG[A/G]CTGGAGTAAGAGCCGAGCACCGGCG
Phenotype
MIM: 602532
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RNF217 PubMed Links

Gene Details

Gene
RNF217
Gene Name
ring finger protein 217
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286398.2 365 Missense Mutation GAC,GGC D77G NP_001273327.1
NM_152553.4 365 Intron NP_689766.1
XM_011535494.2 365 Missense Mutation GAC,GGC D77G XP_011533796.1
XM_011535495.2 365 Missense Mutation GAC,GGC D77G XP_011533797.1
XM_011535496.2 365 Missense Mutation GAC,GGC D77G XP_011533798.1
XM_011535499.2 365 Missense Mutation GAC,GGC D77G XP_011533801.1
XM_011535500.2 365 Intron XP_011533802.1
XM_017010317.1 365 Missense Mutation GAC,GGC D77G XP_016865806.1
Gene
RNF217-AS1
Gene Name
RNF217 antisense RNA 1 (head to head)
There are no transcripts associated with this gene.

View Full Product Details