Product Details

SNP ID
rs185233039
Assay Type
Functionally tested
NCBI dbSNP Submissions
6
Location
Chr.1:108717781 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAACTCTAAAACCTCTTTCAGCTCC[G/T]GAAATACCCACTATTGATCAGGCAT
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
FNDC7 PubMed Links

Gene Details

Gene
FNDC7
Gene Name
fibronectin type III domain containing 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001144937.1 87 Silent Mutation CCG,CCT P29P NP_001138409.1

View Full Product Details