Product Details

SNP ID
rs185329789
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.17:744947 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTCAGCAATGGACTTTAAGAAGCG[C/T]TGCTCGTGTGCCCTCTGGAGCAAGG
Phenotype
MIM: 611627 MIM: 606969
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
DBIL5P PubMed Links
Additional Information
For this assay, SNP(s) [rs7813] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
DBIL5P
Gene Name
diazepam binding inhibitor-like 5, pseudogene
There are no transcripts associated with this gene.

Gene
FAM57A
Gene Name
family with sequence similarity 57 member A
There are no transcripts associated with this gene.

Gene
GEMIN4
Gene Name
gem nuclear organelle associated protein 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015721.2 3433 Silent Mutation CAA,CAG Q1032Q NP_056536.2
XM_005256667.4 3433 Silent Mutation CAA,CAG Q1036Q XP_005256724.1
XM_005256670.4 3433 Silent Mutation CAA,CAG Q1021Q XP_005256727.1
XM_011523910.2 3433 Silent Mutation CAA,CAG Q1036Q XP_011522212.1
XM_011523911.2 3433 Silent Mutation CAA,CAG Q1036Q XP_011522213.1
XM_011523912.2 3433 Silent Mutation CAA,CAG Q1021Q XP_011522214.1
XM_011523913.2 3433 Silent Mutation CAA,CAG Q1021Q XP_011522215.1
XM_017024709.1 3433 Silent Mutation CAA,CAG Q1036Q XP_016880198.1

View Full Product Details