Product Details

SNP ID
rs185283574
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:27356741 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTGACCTCGGTGAGACCGCCTCCT[C/T]CTCCACCTCTGCAGCCCAGTGCGGT
Phenotype
MIM: 604468 MIM: 608042
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MAP3K6 PubMed Links

Gene Details

Gene
MAP3K6
Gene Name
mitogen-activated protein kinase kinase kinase 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001297609.1 3618 Missense Mutation AAG,GAG K1117E NP_001284538.1
NM_004672.4 3618 Missense Mutation AAG,GAG K1125E NP_004663.3
XM_017002771.1 3618 Missense Mutation AAG,GAG K706E XP_016858260.1
XM_017002772.1 3618 Intron XP_016858261.1
Gene
SYTL1
Gene Name
synaptotagmin like 1
There are no transcripts associated with this gene.

View Full Product Details