Product Details

SNP ID
rs185396411
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:17515494 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TACTAATTTCATGAAACATACGAAG[A/T]TCTTCAAGTACTAAGTCAAATTTTC
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
RAD51AP2 PubMed Links

Gene Details

Gene
RAD51AP2
Gene Name
RAD51 associated protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001099218.2 2946 Missense Mutation GAA,GAT E974D NP_001092688.1
NM_001321233.1 2946 Missense Mutation GAA,GAT E965D NP_001308162.1
XM_011533084.1 2946 Missense Mutation GAA,GAT E974D XP_011531386.1

View Full Product Details