Product Details

SNP ID
rs185078297
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:82362216 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTCCTACCTCTACGCCTCCTGGAT[A/G]TATCAGCTTCAACATGGAGATCAGC
Phenotype
MIM: 615647
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TEX19 PubMed Links

Gene Details

Gene
TEX19
Gene Name
testis expressed 19
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_207459.3 463 Missense Mutation ATA,ATG I22M NP_997342.1
XM_017024638.1 463 Missense Mutation ATA,ATG I22M XP_016880127.1
XM_017024639.1 463 Missense Mutation ATA,ATG I22M XP_016880128.1

View Full Product Details