Product Details

SNP ID
rs185081958
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:102838608 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTGTTCTTTTCCTTATCAGAAATG[A/G]CTGCATCGATTTTCCTCACGGTTGG
Phenotype
MIM: 185250
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MMP3 PubMed Links

Gene Details

Gene
MMP3
Gene Name
matrix metallopeptidase 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002422.4 1315 Missense Mutation GCC,GTC A391V NP_002413.1
Gene
WTAPP1
Gene Name
Wilms tumor 1 associated protein pseudogene 1
There are no transcripts associated with this gene.

View Full Product Details