Product Details

SNP ID
rs185137295
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:118898554 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGGCCCATGAGGGAGCCCTGCGGG[C/G]AGAGCATGTGGGGCGGCTGGCTGTA
Phenotype
MIM: 609004 MIM: 601613
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
BCL9L PubMed Links

Gene Details

Gene
BCL9L
Gene Name
B-cell CLL/lymphoma 9-like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_182557.2 5326 Missense Mutation TCC,TGC S1454C NP_872363.1
XM_005271511.2 5326 Missense Mutation TCC,TGC S1454C XP_005271568.1
XM_005271512.2 5326 Missense Mutation TCC,TGC S1417C XP_005271569.1
XM_005271513.2 5326 Missense Mutation TCC,TGC S1417C XP_005271570.1
XM_006718815.2 5326 Missense Mutation TCC,TGC S1454C XP_006718878.1
XM_011542756.2 5326 Missense Mutation TCC,TGC S1469C XP_011541058.1
XM_017017572.1 5326 Missense Mutation TCC,TGC S1502C XP_016873061.1
XM_017017573.1 5326 Missense Mutation TCC,TGC S1499C XP_016873062.1
XM_017017574.1 5326 Missense Mutation TCC,TGC S1468C XP_016873063.1
Gene
CXCR5
Gene Name
C-X-C motif chemokine receptor 5
There are no transcripts associated with this gene.

Gene
LOC101929133
Gene Name
uncharacterized LOC101929133
There are no transcripts associated with this gene.

View Full Product Details