Product Details

SNP ID
rs185696400
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:152649320 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTATGGACAAGCAGGCCTGCAACCT[G/T]GTGGTAGAGAGCTGTACGTATGTCT
Phenotype
MIM: 300349
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
GABRQ PubMed Links

Gene Details

Gene
GABRQ
Gene Name
gamma-aminobutyric acid type A receptor theta subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018558.3 617 Silent Mutation CTG,CTT L199L NP_061028.3
XM_011531184.2 617 Silent Mutation CTG,CTT L199L XP_011529486.1

View Full Product Details