Product Details

SNP ID
rs186020952
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:15563448 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTAGGCTCTGCCTCAGGGATTTTTG[A/G]AGTTCTTCTGGTGTTTGTACAAAAT
Phenotype
MIM: 604300
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HACL1 PubMed Links

Gene Details

Gene
HACL1
Gene Name
2-hydroxyacyl-CoA lyase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001284413.1 2003 Silent Mutation CTC,CTT L511L NP_001271342.1
NM_001284415.1 2003 Silent Mutation CTC,CTT L478L NP_001271344.1
NM_001284416.1 2003 Silent Mutation CTC,CTT L456L NP_001271345.1
NM_012260.3 2003 Silent Mutation CTC,CTT L538L NP_036392.2

View Full Product Details