Product Details

SNP ID
rs186111476
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:100629985 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAATTCACAGGGGGGACCCTAAATG[C/T]CCTAAAATTAAAGAAAAAAAGTTAA
Phenotype
MIM: 300191
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TSPAN6 PubMed Links

Gene Details

Gene
TSPAN6
Gene Name
tetraspanin 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278740.1 1234 UTR 3 NP_001265669.1
NM_001278741.1 1234 UTR 3 NP_001265670.1
NM_001278742.1 1234 Missense Mutation GAC,GGC D136G NP_001265671.1
NM_001278743.1 1234 UTR 3 NP_001265672.1
NM_003270.3 1234 UTR 3 NP_003261.1
XM_011531018.1 1234 UTR 3 XP_011529320.1

View Full Product Details