Product Details

SNP ID
rs185792702
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:40198487 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGACCACCAAGATGAGCGCTGCGGG[G/T]ACCTACGCCTGGATGGCGCCGGAGG
Phenotype
MIM: 600137
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
MAP3K10 PubMed Links

Gene Details

Gene
MAP3K10
Gene Name
mitogen-activated protein kinase kinase kinase 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002446.3 1083 Silent Mutation GGG,GGT G265G NP_002437.2
XM_006723222.2 1083 UTR 5 XP_006723285.1
XM_006723223.3 1083 Intron XP_006723286.1
XM_011526981.2 1083 Silent Mutation GGG,GGT G265G XP_011525283.1
XM_011526982.2 1083 Silent Mutation GGG,GGT G265G XP_011525284.1

View Full Product Details