Product Details
- SNP ID
-
rs185844421
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:126424865 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCCCGTAGTCGTAGAGGCGGCCCTG[A/G]CCGCTCAGGGTGCTGTAGATGTTGG
- Phenotype
-
MIM: 607761
MIM: 104240
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
KIRREL3
PubMed Links
Gene Details
- Gene
- KIRREL3
- Gene Name
- kin of IRRE like 3 (Drosophila)
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001161707.1 |
2232 |
Intron |
|
|
NP_001155179.1 |
NM_001301097.1 |
2232 |
Silent Mutation |
GGC,GGT |
G672G |
NP_001288026.1 |
NM_032531.3 |
2232 |
Silent Mutation |
GGC,GGT |
G684G |
NP_115920.1 |
XM_011543026.2 |
2232 |
Silent Mutation |
GGC,GGT |
G715G |
XP_011541328.1 |
XM_011543027.2 |
2232 |
Silent Mutation |
GGC,GGT |
G709G |
XP_011541329.1 |
XM_011543028.2 |
2232 |
Silent Mutation |
GGC,GGT |
G709G |
XP_011541330.1 |
XM_011543030.2 |
2232 |
Silent Mutation |
GGC,GGT |
G690G |
XP_011541332.1 |
XM_011543031.2 |
2232 |
Silent Mutation |
GGC,GGT |
G684G |
XP_011541333.1 |
XM_011543032.2 |
2232 |
Silent Mutation |
GGC,GGT |
G678G |
XP_011541334.1 |
XM_011543033.1 |
2232 |
Silent Mutation |
GGC,GGT |
G672G |
XP_011541335.1 |
XM_017018418.1 |
2232 |
Silent Mutation |
GGC,GGT |
G709G |
XP_016873907.1 |
XM_017018419.1 |
2232 |
Silent Mutation |
GGC,GGT |
G697G |
XP_016873908.1 |
XM_017018420.1 |
2232 |
Silent Mutation |
GGC,GGT |
G684G |
XP_016873909.1 |
- Gene
- ST3GAL4
- Gene Name
- ST3 beta-galactoside alpha-2,3-sialyltransferase 4
View Full Product Details