Product Details

SNP ID
rs186270963
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:70234058 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGAATTAGCTTCAGGGTTTTTTGT[C/G]TTCCCTTACCCATATCCATTTCGCC
Phenotype
MIM: 601695 MIM: 607241
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CSN3 PubMed Links

Gene Details

Gene
CSN3
Gene Name
casein kappa
There are no transcripts associated with this gene.

Gene
FDCSP
Gene Name
follicular dendritic cell secreted protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152997.3 241 Silent Mutation GTC,GTG V43V NP_694542.1

View Full Product Details