Product Details

SNP ID
rs186291453
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:6240112 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGTGGTGGACAAGGGTAGGATCTC[A/G]AGTCGCTACGTTCGCACCTGGAGTT
Phenotype
MIM: 609472
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CNGA4 PubMed Links

Gene Details

Gene
CNGA4
Gene Name
cyclic nucleotide gated channel alpha 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001037329.3 340 Silent Mutation TCA,TCG S106S NP_001032406.1
XM_017017217.1 340 Silent Mutation TCA,TCG S106S XP_016872706.1
XM_017017218.1 340 Silent Mutation TCA,TCG S66S XP_016872707.1
XM_017017219.1 340 Silent Mutation TCA,TCG S66S XP_016872708.1
Gene
FAM160A2
Gene Name
family with sequence similarity 160 member A2
There are no transcripts associated with this gene.

View Full Product Details