Product Details

SNP ID
rs186456038
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:49689428 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAATGGATTGCCTACCTTCCTCAGG[C/T]GTTTTCACCTCCTCTGGATTTGGCA
Phenotype
MIM: 300288
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PAGE1 PubMed Links

Gene Details

Gene
PAGE1
Gene Name
PAGE family member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003785.3 Intron NP_003776.2
XM_011543998.2 Intron XP_011542300.1

View Full Product Details