Product Details

SNP ID
rs186800052
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:5389541 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCATCCCCTTTTTCTTTATGTACA[A/T]GGTTGCCATCTCAGGCAATTGTTTC
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
OR51B5 PubMed Links

Gene Details

Gene
OR51B5
Gene Name
olfactory receptor family 51 subfamily B member 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005567.2 165 Intron NP_001005567.2
Gene
OR51M1
Gene Name
olfactory receptor family 51 subfamily M member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004756.2 165 Missense Mutation AAG,ATG K48M NP_001004756.2

View Full Product Details