Product Details

SNP ID
rs186821678
Assay Type
Functionally tested
NCBI dbSNP Submissions
10
Location
Chr.1:209432256 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGGAGCTGACAACCATGAGGCCTC[G/T]GCAGCCACCGCCACCACCGCCGCCG
Phenotype
MIM: 613147
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
MIR205 PubMed Links

Gene Details

Gene
MIR205
Gene Name
microRNA 205
There are no transcripts associated with this gene.

Gene
MIR205HG
Gene Name
MIR205 host gene
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001104548.1 599 Silent Mutation TCG,TCT S72S NP_001098018.1
XM_017002058.1 599 Silent Mutation TCG,TCT S72S XP_016857547.1

View Full Product Details