Product Details

SNP ID
rs187074319
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:10577225 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAAAGACTTAATACTCCAAATCAC[A/G]ACGTTTCTCTCCGGCACATACTTGG
Phenotype
MIM: 607309 MIM: 600927
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
AP1M2 PubMed Links

Gene Details

Gene
AP1M2
Gene Name
adaptor related protein complex 1 mu 2 subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001300887.1 1104 Silent Mutation GTC,GTT V342V NP_001287816.1
NM_005498.4 1104 Silent Mutation GTC,GTT V340V NP_005489.2
Gene
CDKN2D
Gene Name
cyclin dependent kinase inhibitor 2D
There are no transcripts associated with this gene.

View Full Product Details