Product Details

SNP ID
rs187060721
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:105110773 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATAGGTTTCTTCTGCTTTATAGAC[C/T]CTGATCCTGAAAATCCAGAACCCAC
Phenotype
MIM: 608444 MIM: 602980
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KMT2E PubMed Links

Gene Details

Gene
KMT2E
Gene Name
lysine methyltransferase 2E
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018682.3 4426 Missense Mutation CCT,TCT P1325S NP_061152.3
NM_182931.2 4426 Missense Mutation CCT,TCT P1325S NP_891847.1
XM_005250493.1 4426 Missense Mutation CCT,TCT P1325S XP_005250550.1
XM_011516400.1 4426 Missense Mutation CCT,TCT P1325S XP_011514702.1
XM_017012435.1 4426 Missense Mutation CCT,TCT P1179S XP_016867924.1
Gene
SRPK2
Gene Name
SRSF protein kinase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278273.1 4426 Intron NP_001265202.1
NM_182691.2 4426 Intron NP_872633.1
NM_182692.2 4426 Intron NP_872634.1
XM_005250549.3 4426 Intron XP_005250606.2
XM_005250550.3 4426 Intron XP_005250607.1
XM_005250551.4 4426 Intron XP_005250608.1
XM_006716098.2 4426 Intron XP_006716161.1
XM_011516536.2 4426 Intron XP_011514838.1
XM_011516538.2 4426 Intron XP_011514840.1
XM_017012560.1 4426 Intron XP_016868049.1
XM_017012561.1 4426 Intron XP_016868050.1
XM_017012562.1 4426 Intron XP_016868051.1
XM_017012563.1 4426 Intron XP_016868052.1
XM_017012564.1 4426 Intron XP_016868053.1
XM_017012565.1 4426 Intron XP_016868054.1
XM_017012566.1 4426 Intron XP_016868055.1

View Full Product Details