Product Details

SNP ID
rs187064013
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:42758189 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCATCCACCACAGGTAGCTTGCGTC[C/T]GGAGTCTCAGGATCACAGGTTAAGC
Phenotype
MIM: 176397
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PSG8 PubMed Links

Gene Details

Gene
PSG8
Gene Name
pregnancy specific beta-1-glycoprotein 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001130167.1 620 Silent Mutation CCA,CCG P174P NP_001123639.1
NM_001130168.1 620 Silent Mutation CCA,CCG P52P NP_001123640.1
NM_182707.2 620 Silent Mutation CCA,CCG P174P NP_874366.1
XM_011526986.1 620 Intron XP_011525288.1
XM_011526987.2 620 Intron XP_011525289.1
XM_011526988.2 620 Intron XP_011525290.1

View Full Product Details