Product Details

SNP ID
rs187244445
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:196722470 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGCATTGAGGACTTGCACACCTGC[C/T]GTCTCTTAATTAAACAGGACATTCC
Phenotype
MIM: 610276
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PIGX PubMed Links

Gene Details

Gene
PIGX
Gene Name
phosphatidylinositol glycan anchor biosynthesis class X
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001166304.1 391 Missense Mutation CGT,TGT R78C NP_001159776.1
NM_017861.3 391 Missense Mutation CGT,TGT R78C NP_060331.3

View Full Product Details