Product Details
- SNP ID
-
rs187227683
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.20:45512795 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCGTCATGTAGGTTTGACAAGGGCC[C/T]TTTTCCATAGGAAAAGCGCATACAT
- Phenotype
-
MIM: 613941
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
SPINT3
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs16990288] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- SPINT3
- Gene Name
- serine peptidase inhibitor, Kunitz type 3
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_006652.1 |
142 |
Silent Mutation |
|
|
NP_006643.1 |
View Full Product Details