Product Details

SNP ID
rs187170506
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:70927302 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCACCTGAGGATGAGAACACAAAT[C/T]GACACCAGGGAGTAAGCAAGCAGGG
Phenotype
MIM: 300443
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SLC7A3 PubMed Links

Gene Details

Gene
SLC7A3
Gene Name
solute carrier family 7 member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001048164.2 1440 Silent Mutation TCA,TCG S422S NP_001041629.1
NM_032803.5 1440 Silent Mutation TCA,TCG S422S NP_116192.4
XM_017029912.1 1440 Silent Mutation TCA,TCG S422S XP_016885401.1

View Full Product Details