Product Details

SNP ID
rs186911920
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:154410432 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGCATGGACAAAGGCCGTGGCTTG[C/T]CTTCAGAGGGACGAGAAAGAAGGTG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CNKSR3 PubMed Links

Gene Details

Gene
CNKSR3
Gene Name
CNKSR family member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_173515.2 4268 Missense Mutation GAC,GGC D427G NP_775786.2
XM_011535485.2 4268 Missense Mutation GAC,GGC D417G XP_011533787.1

View Full Product Details