Product Details

SNP ID
rs186970426
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:75914851 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTACTCAGCTTTTGCTGCGCGGAGA[A/G]CTCCGCCCACTCGGCAGCCAGGCGC
Phenotype
MIM: 616807 MIM: 611844
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FBF1 PubMed Links

Gene Details

Gene
FBF1
Gene Name
Fas binding factor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001319193.1 2793 Missense Mutation CTC,TTC L904F NP_001306122.1
Gene
MRPL38
Gene Name
mitochondrial ribosomal protein L38
There are no transcripts associated with this gene.

View Full Product Details