Product Details

SNP ID
rs186775636
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:155479837 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCAGAGTGCTGAGTGCTTTAACTC[A/G]ATAGTCATCATATGAATTTCAATTG
Phenotype
MIM: 612835
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PLCH1 PubMed Links

Gene Details

Gene
PLCH1
Gene Name
phospholipase C eta 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001130960.1 Intron NP_001124432.1
NM_001130961.1 Intron NP_001124433.1
NM_014996.2 Intron NP_055811.1
XM_005247238.1 Intron XP_005247295.1
XM_005247239.1 Intron XP_005247296.1
XM_011512560.2 Intron XP_011510862.1
XM_011512561.2 Intron XP_011510863.1
XM_011512562.2 Intron XP_011510864.1
XM_011512564.2 Intron XP_011510866.1
XM_011512565.2 Intron XP_011510867.1
XM_011512566.2 Intron XP_011510868.1
XM_011512567.1 Intron XP_011510869.1
XM_017005923.1 Intron XP_016861412.1
XM_017005924.1 Intron XP_016861413.1
XM_017005925.1 Intron XP_016861414.1
XM_017005926.1 Intron XP_016861415.1
XM_017005927.1 Intron XP_016861416.1
XM_017005928.1 Intron XP_016861417.1
XM_017005929.1 Intron XP_016861418.1

View Full Product Details