Product Details

SNP ID
rs187629537
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:57779180 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGTCCAGTCTCATTAACCACAGGA[A/G]AGTTCACTCTGGAGCAAAGCGTTAT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF586 PubMed Links

Gene Details

Gene
ZNF586
Gene Name
zinc finger protein 586
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001077426.2 784 Missense Mutation AAG,GAG K156E NP_001070894.1
NM_001204814.1 784 Missense Mutation AAA,AGA K155R NP_001191743.1
NM_017652.3 784 Missense Mutation AAA,AGA K198R NP_060122.2

View Full Product Details