Product Details

SNP ID
rs187679472
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:30926871 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCTCCGTGTGAAGGCCAAGTTCGA[A/G]GCCAGAGTCCCAGCTTTCTACTACA
Phenotype
MIM: 608635
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ADAP2 PubMed Links

Gene Details

Gene
ADAP2
Gene Name
ArfGAP with dual PH domains 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018404.2 549 Silent Mutation GAA,GAG E90E NP_060874.1

View Full Product Details