Product Details

SNP ID
rs187452429
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:25677914 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTCTCTAGCCCTCCGGTTCGTGCT[C/G]GCCTCTTCAGGGACCCGGCGTGCCT
Phenotype
MIM: 610529
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
TUSC1 PubMed Links

Gene Details

Gene
TUSC1
Gene Name
tumor suppressor candidate 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004125.2 945 Silent Mutation GCC,GCG A136A NP_001004125.1

View Full Product Details