Product Details

SNP ID
rs187402322
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:65579490 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCATAAGCGGCATACTCAGAGGTG[C/G]CGGTACTGGCCAGCTTGAGCTGCTG
Phenotype
MIM: 615940
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
HACD3 PubMed Links

Gene Details

Gene
HACD3
Gene Name
3-hydroxyacyl-CoA dehydratase 3
There are no transcripts associated with this gene.

Gene
VWA9
Gene Name
von Willebrand factor A domain containing 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001136043.2 1730 Missense Mutation GCC,GGC A413G NP_001129515.1
NM_001207058.1 1730 Missense Mutation GCC,GGC A474G NP_001193987.1
NM_001207059.1 1730 Missense Mutation GCC,GGC A435G NP_001193988.1
XM_006720708.3 1730 Missense Mutation GCC,GGC A524G XP_006720771.1
XM_006720709.3 1730 Missense Mutation GCC,GGC A413G XP_006720772.1
XM_006720711.2 1730 Missense Mutation GCC,GGC A413G XP_006720774.1
XM_011522101.2 1730 Missense Mutation GCC,GGC A413G XP_011520403.1
XM_017022644.1 1730 Missense Mutation GCC,GGC A413G XP_016878133.1

View Full Product Details