Product Details

SNP ID
rs188054107
Assay Type
Functionally tested
NCBI dbSNP Submissions
15
Location
Chr.1:156738523 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTACCCCCTGTATGAGGCTCTCACC[A/G]TGTCCCCACAGAACAGATACCAGTC
Phenotype
MIM: 600339 MIM: 611930 MIM: 611836
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HDGF PubMed Links

Gene Details

Gene
HDGF
Gene Name
hepatoma-derived growth factor
There are no transcripts associated with this gene.

Gene
ISG20L2
Gene Name
interferon stimulated exonuclease gene 20 like 2
There are no transcripts associated with this gene.

Gene
MRPL24
Gene Name
mitochondrial ribosomal protein L24
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024540.3 432 Missense Mutation NP_078816.2
NM_145729.2 432 Missense Mutation NP_663781.1
XM_011509981.2 432 Missense Mutation XP_011508283.1
XM_011509982.2 432 Missense Mutation XP_011508284.1
Gene
RRNAD1
Gene Name
ribosomal RNA adenine dimethylase domain containing 1
There are no transcripts associated with this gene.

View Full Product Details