Product Details

SNP ID
rs188050006
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:39727927 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCCTCCTGCCTTCAGCCCAGCCTT[C/T]GACAACCTCTATTACTGGGACCAGG
Phenotype
MIM: 164870 MIM: 601522 MIM: 611802
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ERBB2 PubMed Links

Gene Details

Gene
ERBB2
Gene Name
erb-b2 receptor tyrosine kinase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005862.2 4137 Silent Mutation TTC,TTT F1187F NP_001005862.1
NM_001289936.1 4137 Silent Mutation TTC,TTT F1202F NP_001276865.1
NM_001289937.1 4137 UTR 3 NP_001276866.1
NM_001289938.1 4137 Intron NP_001276867.1
NM_004448.3 4137 Silent Mutation TTC,TTT F1217F NP_004439.2
Gene
GRB7
Gene Name
growth factor receptor bound protein 7
There are no transcripts associated with this gene.

Gene
MIEN1
Gene Name
migration and invasion enhancer 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032339.4 4137 Intron NP_115715.3
XM_005257736.3 4137 Intron XP_005257793.1
Gene
MIR4728
Gene Name
microRNA 4728
There are no transcripts associated with this gene.

View Full Product Details