Product Details

SNP ID
rs188426984
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:56663972 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATCACAAAGTTATCTCCTTCTCCA[G/T]CTGTGCCACCCAGTTTGCTTTTTTT
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
OR5AR1 PubMed Links

Gene Details

Gene
OR5AR1
Gene Name
olfactory receptor family 5 subfamily AR member 1 (gene/pseudogene)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004730.1 287 Missense Mutation AGC,ATC S96I NP_001004730.1

View Full Product Details