Product Details

SNP ID
rs188852690
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:110468484 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAAGCTTGGATACCTGAGTAATAAC[C/G]GACAACAGATAAAGCTCCGCATCCT
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
FAM216A PubMed Links

Gene Details

Gene
FAM216A
Gene Name
family with sequence similarity 216 member A
There are no transcripts associated with this gene.

Gene
GPN3
Gene Name
GPN-loop GTPase 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001164372.1 352 Silent Mutation TCC,TCG S51S NP_001157844.1
NM_001164373.1 352 Intron NP_001157845.1
NM_016301.3 352 Intron NP_057385.3
XM_005253896.4 352 Missense Mutation CCG,CGG P112R XP_005253953.3
XM_017019394.1 352 Intron XP_016874883.1

View Full Product Details