Product Details

SNP ID
rs188824106
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:30570244 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCATCCATGCTGGGTATGGGTGCC[C/G]GGGGCTGTGCTCCTGTGGTCGGGTC
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ZNF688 PubMed Links

Gene Details

Gene
ZNF688
Gene Name
zinc finger protein 688
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001024683.1 593 Missense Mutation CCG,CGG P154R NP_001019854.1
NM_145271.3 593 Missense Mutation CCG,CGG P168R NP_660314.1
XM_005255139.2 593 Missense Mutation CCG,CGG P91R XP_005255196.1
XM_005255140.2 593 Missense Mutation CCG,CGG P91R XP_005255197.1
Gene
ZNF785
Gene Name
zinc finger protein 785
There are no transcripts associated with this gene.

View Full Product Details