Product Details

SNP ID
rs188794826
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:42679981 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCGCGTAGCAGGCTACCATGACGC[C/T]CAGCGGCAGCGCGAAGCCCAGGGCC
Phenotype
MIM: 600240 MIM: 602346
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CCR10 PubMed Links

Gene Details

Gene
CCR10
Gene Name
C-C motif chemokine receptor 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016602.2 19 Missense Mutation AGC,GGC S221G NP_057686.2
Gene
CNTNAP1
Gene Name
contactin associated protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003632.2 19 Intron NP_003623.1
XM_005257748.4 19 Intron XP_005257805.1
XM_017025238.1 19 UTR 5 XP_016880727.1
Gene
PLEKHH3
Gene Name
pleckstrin homology, MyTH4 and FERM domain containing H3
There are no transcripts associated with this gene.

View Full Product Details