Product Details

SNP ID
rs188363199
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:57391372 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGGGGTACACTCGGGCGAGAGGCCC[C/T]CTCACTCCCTCTCTAGTAATGCAAG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NAA30 PubMed Links

Gene Details

Gene
NAA30
Gene Name
N(alpha)-acetyltransferase 30, NatC catalytic subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001011713.2 569 Missense Mutation CCT,TCT P139S NP_001011713.2

View Full Product Details