Product Details

SNP ID
rs188404570
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:67394734 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGTCGGCCCCGGCCCCGCGGGGCG[C/G]CCAGAGCGGCGCTGCCCAGGCGCGT
Phenotype
MIM: 616957
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
LRRC36 PubMed Links

Gene Details

Gene
LRRC36
Gene Name
leucine rich repeat containing 36
There are no transcripts associated with this gene.

Gene
TPPP3
Gene Name
tubulin polymerization promoting protein family member 3
There are no transcripts associated with this gene.

Gene
ZDHHC1
Gene Name
zinc finger DHHC-type containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001323627.1 1304 Missense Mutation GCC,GGC A442G NP_001310556.1
NM_013304.2 1304 Missense Mutation CCG,GCG P464A NP_037436.1
XM_005255915.1 1304 UTR 3 XP_005255972.1
XM_006721185.1 1304 Missense Mutation CCG,GCG P449A XP_006721248.1
XM_011523058.1 1304 Missense Mutation GCC,GGC A429G XP_011521360.1
XM_011523059.1 1304 Missense Mutation CCG,GCG P411A XP_011521361.1
XM_011523060.1 1304 Missense Mutation CCG,GCG P379A XP_011521362.1

View Full Product Details