Product Details

SNP ID
rs189387085
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:73700699 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAAGATCCACCAGCCTGGCCCACA[C/T]GTGGAAGAAGGACTGGCCCGTCTTC
Phenotype
MIM: 608190
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NAT8B PubMed Links

Gene Details

Gene
NAT8B
Gene Name
N-acetyltransferase 8B (putative, gene/pseudogene)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016347.2 642 Missense Mutation ATG,GTG M203V NP_057431.2

View Full Product Details