Product Details
- SNP ID
-
rs189396476
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.9:120394591 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGTTTTGCAACTTCTTTTCTTGTTC[A/C]AACAATTTTTTATGTAGTTTGGTGA
- Phenotype
-
MIM: 608201
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
CDK5RAP2
PubMed Links
Gene Details
- Gene
- CDK5RAP2
- Gene Name
- CDK5 regulatory subunit associated protein 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001011649.2 |
5584 |
Missense Mutation |
TTG,TTT |
L1754F |
NP_001011649.1 |
NM_001272039.1 |
5584 |
Missense Mutation |
TTG,TTT |
L1603F |
NP_001258968.1 |
NM_018249.5 |
5584 |
Missense Mutation |
TTG,TTT |
L1833F |
NP_060719.4 |
XM_006717182.1 |
5584 |
Missense Mutation |
TTG,TTT |
L1801F |
XP_006717245.1 |
XM_006717185.1 |
5584 |
Missense Mutation |
TTG,TTT |
L1604F |
XP_006717248.1 |
XM_011518860.1 |
5584 |
Missense Mutation |
TTG,TTT |
L1832F |
XP_011517162.1 |
XM_017014921.1 |
5584 |
Missense Mutation |
TTG,TTT |
L1800F |
XP_016870410.1 |
XM_017014922.1 |
5584 |
Missense Mutation |
TTG,TTT |
L1555F |
XP_016870411.1 |
XM_017014923.1 |
5584 |
Missense Mutation |
TTG,TTT |
L1525F |
XP_016870412.1 |
XM_017014924.1 |
5584 |
Missense Mutation |
TTG,TTT |
L1019F |
XP_016870413.1 |
View Full Product Details