Product Details

SNP ID
rs189434887
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:1936849 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGGGTGAGTGGTGTTCCTTGCGGG[C/T]GGCCCTGTCGGTGGTGGTGAGCGTG
Phenotype
MIM: 610461
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RTN4RL1 PubMed Links

Gene Details

Gene
RTN4RL1
Gene Name
reticulon 4 receptor like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_178568.3 1453 Missense Mutation NP_848663.1

View Full Product Details